Skip to Content
Merck
All Photos(1)

Key Documents

SRP0131

Sigma-Aldrich

EZH1/EED/SUZ12/RbAp48/AEBP2 human

recombinant, expressed in baculovirus infected insect cells, ≥90% (SDS-PAGE)

Synonym(s):

ENX-2, EZH1, Enhancer of zeste homolog 1, KMT6B, Lysine N-methyltransferase 6B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in baculovirus infected insect cells

Assay

≥90% (SDS-PAGE)

form

aqueous solution

mol wt

48 kDa (RbAp48)
51 kDa (EED)
53 kDa (AEBP2)
86 kDa (EZH1)
87 kDa (SUZ12)

packaging

pkg of 50 μg

concentration

>0.02 mg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... EZH1(2145)

General description

Complex of: Human EZH1, GenBank Accession No. NM_001991, amino acids 2-end, with N-terminal His tag, MW = 86 kDa. Human EED, NM_003797, amino acids 2-end, with N-terminal DDDDK tag (FLAG), MW = 51 kDa. Human SUZ12, NM_015355, amino acids 2-end, with N-terminal His tag. RbAp48 and AEBP2.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Unit Definition

One unit is defined as the amount of enzyme required to methylate 1 pmol of substrate/min at 37°C.

Physical form

Formulated in 25 mM Tris-HCl, pH 8.0, 138 mM NaCl, 0.05% Tween-20 and 10% glycerol.

Preparation Note

Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service