Skip to Content
Merck
All Photos(1)

Documents

SML1214

Sigma-Aldrich

CZC24832

≥98% (HPLC)

Synonym(s):

5-(2-Amino-8-fluoro-[1,2,4]triazolo[1,5-a]pyridin-6-yl)-N-tert-butylpyridine-3-sulfonamide, 5-(2-Amino-8-fluoro[1,2,4]triazolo[1,5-a]pyridin-6-yl)-N-(1,1-dimethylethyl)-3-pyridinesulfonamide, CZC-24832

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H17FN6O2S
CAS Number:
Molecular Weight:
364.40
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

−20°C

SMILES string

FC1=CC(C2=CN=CC(S(NC(C)(C)C)(=O)=O)=C2)=CN3C1=NC(N)=N3

InChI

1S/C15H17FN6O2S/c1-15(2,3)21-25(23,24)11-4-9(6-18-7-11)10-5-12(16)13-19-14(17)20-22(13)8-10/h4-8,21H,1-3H3,(H2,17,20)

InChI key

RXRZPHQBTHQXSV-UHFFFAOYSA-N

Biochem/physiol Actions

CZC24832 is a potent and selective inhibitor of phosphoinositide 3-kinase γ (PI3Kγ) with Kd app of 19 nM. CZC24832 is said to be the first selective PI3Kγ inhibitor with efficacy in in vitro and in vivo models of inflammation. CZC24832 strongly inhibited T-cell interleukin 17A (IL-17A) production by inhibiting expression of retinoic acid receptor–related orphan receptor γt (RORγt), a transcription factor that drives TH17 differentiation, showing a new role for PI3Kγ.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service