SML0760
WIKI4
≥98% (HPLC)
Synonym(s):
2-[3-[[4-(4-Methoxyphenyl)-5-(4-pyridinyl)-4H-1,2,4-triazol-3-yl]thio]propyl]-1Hbenz[de]isoquinoline-1,3(2H)-dione
About This Item
Assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 10 mg/mL, clear (warmed)
storage temp.
−20°C
InChI
1S/C29H23N5O3S/c1-37-22-11-9-21(10-12-22)34-26(20-13-15-30-16-14-20)31-32-29(34)38-18-4-17-33-27(35)23-7-2-5-19-6-3-8-24(25(19)23)28(33)36/h2-3,5-16H,4,17-18H2,1H3
InChI key
RNUXIZKXJOGYQP-UHFFFAOYSA-N
Biochem/physiol Actions
Features and Benefits
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service