SCP0207
PTH Peptide
≥95% (HPLC), lyophilized powder, bovine, 1-34
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C185H293N55O50
Molecular Weight:
4087.64
UNSPSC Code:
12352209
NACRES:
NA.32
Recommended Products
Product Name
[Nle8,18,Tyr34]-PTH 1-34 bovine,
Assay
≥95% (HPLC)
form
lyophilized
composition
Peptide Content, ≥72%
storage condition
protect from light
storage temp.
−20°C
Amino Acid Sequence
Ala-Val-Ser-Glu-Ile-Gln-Phe-Nle-His-Asn-Leu-Gly-Lys-His-Leu-Ser-Ser-Nle-Glu-Arg-Val-Glu-Trp-Leu-Arg-Lys-Lys-Leu-Gln-Asp-Val-His-Asn-Tyr-NH2
Application
Parathyroid hormone (PTH) is an 84 amino acid hormone involved in calcium regulation in counteraction to calcitonin. PTH regulates serum phosphate levels and vitamin D biosynthesis. N-terminal peptides of PTH are used to elicit and study a wide variety of responses in target cells. PTH N-terminal peptides include: PTH (1-34); [Nle8,18,Tyr34]-PTH 1-34; PTH (2-34); [Nle8,18,Tyr34]-PTH (2-34); PTH (3-34); [Nle8,18,Tyr34]-PTH (3-34); PTH (4-34) and PTH (5-34).
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Estelle N Bianchi et al.
Bone, 45(4), 716-725 (2009-06-30)
Interaction of the cytoplasmic adaptor molecule beta-arrestin2 with the activated parathyroid hormone (PTH)/PTHrP receptor inhibits G protein mediated signaling and triggers MAPKs signaling. In turn, the effects of both intermittent (i.) and continuous (c.) PTH on bone are altered in
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service