Skip to Content
Merck
All Photos(1)

Key Documents

SAB2101410

Sigma-Aldrich

Anti-LZTR1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Leucine-zipper-like transcription regulator 1, Anti-MGC21205, Anti-TCFL2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

95 kDa

species reactivity

rabbit, dog, guinea pig, human, bovine, rat, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LZTR1(8216)

Immunogen

Synthetic peptide directed towards the N terminal region of human LZTR1

Biochem/physiol Actions

Leucine-zipper-like transcriptional regulator 1(LZTR1) is belived to be a DNA-binding protein and transcriptional regulator based on its predicted structural characteristics. The transcript is present in several essential fetal organs and is hemizygously deleted in some DiGeorge syndrome patients. LZTR1 is thought to play a critical role in embryogenesis.This gene encodes a member of the BTB-kelch superfamily. Initially described as a putative transcriptional regulator based on weak homology to members of the basic leucine zipper-like family, the encoded protein subsequently has been shown to localize exclusively to the Golgi network where it may help stabilize the Gogli complex. Deletion of this gene may be associated with DiGeorge syndrome.

Sequence

Synthetic peptide located within the following region: AGPGSTGGQIGAAALAGGARSKVAPSVDFDHSCSDSVEYLTLNFGPFETV

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service