Skip to Content
Merck
All Photos(3)

Key Documents

SAB1411332

Sigma-Aldrich

Anti-TCEB3 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

FLJ38760, FLJ42849, SIII, TCEB3A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 87.2 kDa

species reactivity

human

technique(s)

immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TCEB3(6924)

General description

This gene encodes the protein elongin A, which is a subunit of the transcription factor B (SIII) complex. The SIII complex is composed of elongins A/A2, B and C. It activates elongation by RNA polymerase II by suppressing transient pausing of the polymerase at many sites within transcription units. Elongin A functions as the transcriptionally active component of the SIII complex, whereas elongins B and C are regulatory subunits. Elongin A2 is specifically expressed in the testis, and capable of forming a stable complex with elongins B and C. The von Hippel-Lindau tumor suppressor protein binds to elongins B and C, and thereby inhibits transcription elongation. (provided by RefSeq)

Immunogen

TCEB3 (NP_003189.1, 1 a.a. ~ 772 a.a) full-length human protein.

Sequence
MAAESALQVVEKLQARLAANPDPKKLLKYLKKLSTLPITVDILAETGVGKTVNSLRKHEHVGSFARDLVAQWKKLVPVERNAEPDEQDFEKSNSRKRPRDALQKEEEMEGDYQETWKATGSRSYSPDHRQKKHRKLSELERPHKVSHGHERRDERKRCHRMSPTYSSDPESSDYGHVQSPPSCTSPHQMYVDHYRSLEEDQEPIVSHQKPGKGHSNAFQDRLGASQERHLGEPHGKGVVSQNKEHKSSHKDKRPVDAKSDEKASVVSREKSHKALSKEENRRPPSGDNAREKPPSSGVKKEKDREGSSLKKKCLPPSEAASDNHLKKPKHRDPEKAKLDKSKQGLDSFDTGKGAGDLLPKVKEKGSNNLKTPEGKVKTNLDRKSLGSLPKVEETDMEDEFEQPTMSFESYLSYDQPRKKKKKIVKTSATALGDKGLKKNDSKSTGKNLDSVQKLPKVNKTKSEKPAGADLAKLRKVPDVLPVLPDLPLPAIQANYRPLPSLELISSFQPKRKAFSSPQEEEEAGFTGRRMNSKMQVYSGSKCAYLPKMMTLHQQCIRVLKNNIDSIFEVGGVPYSVLEPVLERCTPDQLYRIEEYNHVLIEETDQLWKVHCHRDFKEERPEEYESWREMYLRLQDAREQRLRVLTKNIQFAHANKPKGRQAKMAFVNSVAKPPRDVRRRQEKFGTGGAAVPEKIKIKPAPYPMGSSHASASSISFNPSPEEPAYDGPSTSSAHLAPVVSSTVSYDPRKPTVKKIAPMMAKTIKAFKNRFSRR

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service