Skip to Content
Merck
All Photos(3)

Key Documents

SAB1401761

Sigma-Aldrich

Monoclonal Anti-ENAH antibody produced in mouse

clone 3E6, purified immunoglobulin, buffered aqueous solution

Synonym(s):

ENA, MENA, NDPP1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3E6, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

capture ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ENAH(55740)

Immunogen

ENAH (NP_001008493.1, 291 a.a. ~ 390 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
LQAASQPAETPSQQGIVLGPLAPPPPPPLPPGPAQASVALPPPPGPPPPPPLPSTGPPPPPPPPPLPNQVPPPPPPPPAPPLPASGFFLASMSEDNRPLT

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Harmonization of autoimmune diagnostics with antinuclear antibody testing algorithm: approach of appropriateness and clinical relevance.
Alessandra Melegari et al.
The Israel Medical Association journal : IMAJ, 16(10), 640-642 (2014-12-03)
Thomas E Rohan et al.
Journal of the National Cancer Institute, 106(8) (2014-06-05)
Tumor microenvironment of metastasis (TMEM), consisting of direct contact between a macrophage, an endothelial cell, and a tumor cell, has been associated with metastasis in both rodent mammary tumors and human breast cancer. We prospectively examined the association between TMEM
Ling Lin et al.
Experimental cell research, 327(1), 113-122 (2014-05-27)
Mammalian enabled (MENA), usually known as a direct regulator of microfilament polymerization and bundling, promotes metastasis in various cancers. Here we focus on the role of MENA in hepatocellular carcinoma (HCC) metastasis and the relevant mechanism from the view of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service