SAB1306443
ANTI-S39A3 (CENTER) antibody produced in rabbit
affinity isolated antibody, buffered aqueous solution
Synonym(s):
SLC39A3, Solute carrier family 39 member 3, ZIP3, Zinc transporter ZIP3, Zrt- and Irt-like protein 3
About This Item
biological source
rabbit
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
mol wt
33601 Da
species reactivity
human, mouse
technique(s)
immunohistochemistry: 1:50-1:100
western blot: 1:1000
NCBI accession no.
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... SLC39A3(29985)
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service