S9945
Stemline® Keratinocyte Growth Supplement
With bovine pituitary extract (BPE) and CaCl2, For use with S0196, Stemline® Keratinocyte Medium II
Synonym(s):
Keratinocyte Medium Supplement
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
MDL number:
UNSPSC Code:
12352200
Recommended Products
form
liquid
quality
With bovine pituitary extract (BPE) and CaCl2
manufacturer/tradename
Sigma-Aldrich
color
clear pink-brown
shipped in
dry ice
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
Application
Produced in a GMP state-of-the-art facility, Stemline Keratinocyte Stem Cell Expansion Medium is clearly an excellent choice for your Keratinocyte applications.
Legal Information
Stemline is a registered trademark of Merck KGaA, Darmstadt, Germany
required but not provided
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service