Skip to Content
Merck
All Photos(1)

Key Documents

P0981

Sigma-Aldrich

Anti-Potassium Channel TASK-1 antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human, rat

technique(s)

western blot (chemiluminescent): 1:200

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... KCNK3(3777)
mouse ... Kcnk3(16527)
rat ... Kcnk3(29553)

Related Categories

Immunogen

synthetic peptide corresponding to amino acid residues 252-269 of human TASK-1 (with additional C-terminal cysteine). The epitope is highly conserved in mouse and rat TASK-1.

Physical form

Lyophilized at ~0.6 mg/ml from phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin, 5% sucrose, and 0.025% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sven G Meuth et al.
The Journal of biological chemistry, 283(21), 14559-14570 (2008-04-01)
Two major K(+) channels are expressed in T cells, (i) the voltage-dependent K(V)1.3 channel and (ii) the Ca(2+)-activated K(+) channel KCa 3.1 (IKCa channel). Both critically influence T cell effector functions in vitro and animal models in vivo. Here we
F Duprat et al.
The EMBO journal, 16(17), 5464-5471 (1997-10-06)
TASK is a new member of the recently recognized TWIK K+ channel family. This 395 amino acid polypeptide has four transmembrane segments and two P domains. In adult human, TASK transcripts are found in pancreas<placenta<brain<lung, prostate<heart, kidney<uterus, small intestine and
Alexandra Mant et al.
Channels (Austin, Tex.), 7(4), 288-302 (2013-06-29)
Acid-sensitive, two-pore domain potassium channels, K(2P)3.1 and K(2P)9.1, are implicated in cardiac and nervous tissue responses to hormones, neurotransmitters and drugs. K(2P)3.1 and K(2P)9.1 leak potassium from the cell at rest and directly impact membrane potential. Hence altering channel number
Constanze Schmidt et al.
Circulation, 132(2), 82-92 (2015-05-09)
Antiarrhythmic management of atrial fibrillation (AF) remains a major clinical challenge. Mechanism-based approaches to AF therapy are sought to increase effectiveness and to provide individualized patient care. K(2P)3.1 (TASK-1 [tandem of P domains in a weak inward-rectifying K+ channel-related acid-sensitive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service