Skip to Content
Merck
All Photos(3)

Key Documents

HPA014499

Sigma-Aldrich

Anti-RNF130 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Goliath homolog precursor, Anti-RING finger protein 130

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

PWLSEHCTCPMCKLNILKALGIVPNLPCTDNVAFDMERLTRTQAVNRRSA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RNF130(55819)

General description

RING finger protein130 (RNF130) is a zinc finger protein and contains ring finger motif. RNF130 is a member of Goliath family. It is mapped to human chromosome 5q35.3. It consists of signal peptide sequences at its N-terminus and a unique extracellular or luminal domain. RNF130 alignment is identical with RING, protease-associated (PA), transmembrane (TM) domains. It contains 419 amino acids and is expressed ubiquitously. It is highly expressed in liver and leukocytes.

Immunogen

Goliath homolog precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The human RING finger protein130, also referred to as h-Goliath, functions as a E3 ubiquitin ligase. Development of T cell clonal anergy is regulated by E3 ligase ubiquitination. In hydatidiform mole tissue, RING finger protein130 is upregulated. In Barrett esophagus (BE) and esophageal adenocarcinoma (EAC), RNF130 protein is upregulated in comparison with the normal esophagus.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72845

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Table of Contents

Anti-RNF130 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xianglan Jin et al.
The FEBS journal, 278(1), 78-84 (2010-11-17)
Protein ubiquitination mediated by ubiquitin ligases plays a very important role in a wide spectrum of biological processes including development and disease pathogenesis. RING finger protein 13 (RNF13) is a recently identified ubiquitin ligase which contains an N-terminal protease-associated domain
Seung Jo Kim et al.
Gynecologic oncology, 103(2), 654-660 (2006-06-27)
To gain a better understanding of the genes involved in the pathogenesis of gestational trophoblastic diseases, we evaluated the genome-wide expression levels of genes in complete hydatidiform mole (H-mole) as compared to normal placenta using cDNA microarray technique. The expression
Xing Wei Wang et al.
Gut and liver, 8(5), 487-494 (2014-09-18)
To investigate the differential expression of RING finger (RNF) proteins in Barrett esophagus (BE) and esophageal adenocarcinoma (EAC). The differential expression of RNFs in normal esophagus (NE), BE, and EAC was screened using microarray assay. Real-time quantitative polymerase chain reaction
Nobuhiro Nakamura
Membranes, 1(4), 354-393 (2011-01-01)
A large number of RING finger (RNF) proteins are present in eukaryotic cells and the majority of them are believed to act as E3 ubiquitin ligases. In humans, 49 RNF proteins are predicted to contain transmembrane domains, several of which
T Suami et al.
Carbohydrate research, 53(2), 239-246 (1977-02-01)
The aminocyclitol antibiotic neamine has been modified chemically by removing one or two hydroxyl groups from the 2-deoxystreptamine moiety to give 5- and 6- deoxyneamines (5 and 10), as well as 5,6-dideoxyneamine (15). Their antimicrobial activities were determined against several

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service