Skip to Content
Merck
All Photos(1)

Key Documents

EP6731000341

Eppendorf® CellXpert® C170i CO2 Incubator

new

water/humidity monitoring, Handle on right, inner door with 4 door segments, AC/DC input 220—240 V, 50—60 Hz, EuroPlug

Synonym(s):

Heaters

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104405
NACRES:
NB.02

packaging

pack of 1 ea

manufacturer/tradename

Eppendorf® 6731000341

parameter

±4-50 °C ambient temp. range

W × depth × H

53.9 cm (21.2 in.) × 44.5 cm (14.5 in.) × 69.2 cm (27.3 in.)

platform W × depth

52.2 cm (20.6 in.) ×  42.8 cm (16.9 in.)

volume

167 L

Looking for similar products? Visit Product Comparison Guide

General description

Today′s cell culture requires more than just technical solutions for optimized growth conditions and efficient contamination protection. The CellXpert® CO 2 incubator family includes the first cell culture incubators developed and manufactured by Eppendorf in Hamburg. They are designed to meet the needs of modern industrial and academic cell culture laboratories to reduce time to market or publication with minimal effort and resource use. The development of the CellXpert® family is based, for example, on B. based on decades of knowledge gained in cell culture laboratories from the use of conventional fan-assisted cell culture incubators. These laboratories are the perfect partner for demanding applications such as assay development or the culture of stem cells, primary cells or tissues. Learn more and see more videos and work below. Is protection against laboratory downtime and sample loss essential for you to deliver cell culture results on time? Can′t afford to spend too much time and money on standardized cleaning of your cell culture incubator, on documentation or on replacing expensive, non-sterilizable HEPA filters with a fan connection? The CellXpert′s strongest contamination protection includes, among other things, the fanless design. It effectively reduces the spread of air contaminants, eliminating the need to use internal HEPA filters with a fan connection. order to properly compare cell growth in vessels placed at different locations in the incubator, the temperature must be very In order to properly compare cell growth in vessels placed at different locations in the incubator, the temperature must be very uniform. Together with state-of-the-art microprocessor control, this is achieved in CellXpert® cell culture incubators by replacing the traditional arrangement of one or two temperature sensors (the second is almost always only used to control overtemperature) with multiple independent sensors in different locations. The effectiveness of this innovative approach was verified by measuring the temperature deviation at 27 points inside the incubator (according to DIN DIN 12880), which far exceeds the required specification.uniform. Together with state-of-the-art microprocessor control, this is achieved in CellXpert® cell culture incubators by replacing the traditional arrangement of one or two temperature sensors (the second is almost always only used to control overtemperature) with multiple independent sensors in different locations. The effectiveness of this innovative approach was verified by measuring the temperature deviation at 27 points inside the incubator (according to DIN DIN 12880), which far exceeds the required specification. In general, HEPA filter cartridges are heat sensitive and must be removed before an HTD. Reinstalling the HEPA filter (which capturesIn order to properly compare cell growth in vessels placed at different locations in the incubator, the temperature must be very uniform. Together with state-of-the-art microprocessor control, this is achieved in CellXpert® cell culture incubators by replacing the traditional arrangement of one or two temperature sensors (the second is almost always only used to control overtemperature) with multiple independent sensors in different locations.

Features and Benefits

  • 180 °C high temperature disinfection with clear step-by-step instructions on the screen and implementation protocol for standardized procedures that can be downloaded
  • The few interior parts can be removed
  • from the incubator in 40 s and quickly wiped down
  • No hiding place for Contamination and quick to clean: Smooth incubator chamber with no bumps
  • Fanless design
  • No non-sterilizable HEPA filters with fan attachment that can be potential sources of contamination
  • Options for copper trim and partitioned interior doors for added protection

Legal Information

CellXpert is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service