EP0030120213
Eppendorf® Safe-Lock micro test tubes
capacity 2.0 mL, red, pack of 1000 ea
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121702
NACRES:
NB.11
material
cap (plug seal)
polypropylene cap
polypropylene tube
red
red cap
red tube
round bottom
feature
graduations: no
neck thread Push fit
packaging
pack of 1000 ea
manufacturer/tradename
Eppendorf® 0030120213
capacity
2.0 mL
Looking for similar products? Visit Product Comparison Guide
Features and Benefits
- The hinged Eppendorf® Safe-Lock lid provides additional protection against unintentional lid opening during incubation and storage.
- g-Safe®:Provides great centrifugation stability and prevents sample loss due to tube breakage
- The optimized sealing properties ensures safety when working with hazardous samples and minimal evaporation rates during long-term storage.
- The large frosted lid and wide side-surface area make labeling convenient.
- Functions between temperatures -86 °C to 100 °C
- Autoclavable when open (121 °C, 20 min)
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
g-Safe is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service