Skip to Content
Merck
All Photos(1)

Key Documents

EHU025001

Sigma-Aldrich

MISSION® esiRNA

targeting human CMKLR1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GACTTATCCCCCTTGGAAGCCAGGGTGACCAGGATCTTCCTGGTGGTGGTCTACAGCATCGTCTGCTTCCTCGGGATTCTGGGCAATGGTCTGGTGATCATCATTGCCACCTTCAAGATGAAGAAGACAGTGAACATGGTCTGGTTCCTCAACCTGGCAGTGGCAGATTTCCTGTTCAACGTCTTCCTCCCAATCCATATCACCTATGCCGCCATGGACTACCACTGGGTTTTCGGGACAGCCATGTGCAAGATCAGCAACTTCCTTCTCATCCACAACATGTTCACCAGCGTCTTCCTGCTGACCATCATCAGCTCTGACCGCTGCATCTCTGTGCTCCTCCCTGTCTGGTCCCAGAACCACCGCAGCGTTCGCCTGGCTTACATGGCCTGCATGGTCATCTGGGTCCTGGCTTTCTT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuan Wang et al.
Journal of cellular biochemistry, 120(4), 6459-6470 (2018-11-15)
Psoriasis is a chronic disease which carries the emotional and social burden, promotes joint disability and raises comorbidity possibility in patients. Obesity is closely correlated with the occurrence of psoriasis and adipokines produced by adipose tissues were found to be
Yixin Zhang et al.
Brain, behavior, and immunity, 70, 179-193 (2018-03-03)
Chemerin, an adipokine, has been reported to reduce the production of pro-inflammatory cytokines and neutrophil infiltration. This study investigated the role of Chemerin and its natural receptor, ChemR23, as well as its downstream mediator calmodulin-dependent protein kinase kinase 2 (CAMKK2)/adenosine
Yixin Zhang et al.
Cell death & disease, 10(2), 97-97 (2019-02-06)
Hypoxic-ischemic encephalopathy (HIE) is a devastating neurological event that contributes to the prolonged neurodevelopmental consequences in infants. Therapeutic strategies focused on attenuating neuronal apoptosis in the penumbra appears to be promising. Given the increasingly recognized neuroprotective roles of adipokines in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service