Skip to Content
Merck
All Photos(1)

Key Documents

CLS3262

Corning® Ultra-low attachment culture dishes

100mm petri dish, low-attachment

Synonym(s):

cell culture flasks, tissue culture flasks

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121812
NACRES:
NB.13

material

polystyrene
round

sterility

sterile

packaging

case of 20

manufacturer/tradename

Corning 3262

D × H

100 mm × 20 mm

surface area

55 cm2

color

clear

Looking for similar products? Visit Product Comparison Guide

General description

  • Ultra Low Attachment dishes feature a covalently bound hydrogel layer that effectively inhibits cellular attachment
  • Surface minimizes protein absorption, enzyme activation, and cellular activation
  • Surface is non-cytotoxic, biologically inert, and nondegradable
  • Sterilized by gamma irradiation and certified nonpyrogenic
  • Actual inside growth surface diameter is 80.5mm
  • Height is 20mm
  • Manufactured from optically-clear virgin polystyrene
  • Have stacking beads to aid in handling
  • Supplied with vents to provide consistent gas exchange

Legal Information

Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service