Skip to Content
Merck
All Photos(1)

Key Documents

H357

6092004, human mouth, Polygonal

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

H357, 06092004

biological source

human mouth

growth mode

Adherent

karyotype

Not specified

morphology

Polygonal

products

Not specified

receptors

TGFbeta type I receptor, TGFbeta II receptor, TGFbeta III receptor

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human oral squamous cell carcinoma, tongue

Cell Line Description

Established from a squamous cell carcinoma (SCC) of the tongue (20mm) of a 74 year-old male patient. STNMP stage I, well differentiated, node negative tumour. Mutant p53, codon 110 exon 4, G to A; the previously reported mutant Ha-ras status of this cell line: codon: 13, G to A, codon 61, A-G is under investigation. This cell line is highly responsive to TGF (Transforming growth factor )-β and undergoes epithelial to mesenchymal transition. It expresses high levels of TGF-β1. Fahey et al (1996). Tumourigenic in athymic nude mice on subcutaneous injection and non-tumorigenic on orthotopic injection. Haplotype information: A*02,A*31; B*40,B*44; Cw*03,Cw*05

Culture Medium

DMEM:HAMS F12 (1:1) + 2mM Glutamine + 10% Foetal Bovine Serum (FBS) + 0.5 ug/ml sodium hydrocortisone succinate

Subculture Routine

Split sub-confluent cultures (70-80%), approximately every 5-6 days, 1:8 to 1:10 using 0.05% trypsin/EDTA; 5% CO2; 37°C. Suggested seeding density 5 x 1000 cells/cm2. Cells can take approximately 10 minutes to detach, an alternative is to trypsinise 2

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service