Skip to Content
Merck
All Photos(4)

Key Documents

AV53559

Sigma-Aldrich

Anti-CRAT antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-CAT1, Anti-Carnitine acetyltransferase, Anti-RP11-247A12.5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

51 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CRAT(1384)

Immunogen

Synthetic peptide directed towards the N terminal region of human CRAT

Biochem/physiol Actions

Carnitine acetyltransferase (CRAT) is a key enzyme in the metabolic pathway in mitochondria, peroxisomes and endoplasmic reticulum. CRAT catalyzes the reversible transfer of acyl groups from an acyl-CoA thioester to carnitine and regulates the ratio of acylCoA/CoA in the subcellular compartments. Different subcellular localizations of the CRAT mRNAs are thought to result from alternative splicing of the CRAT gene suggested by the divergent sequences in the 5′ region of peroxisomal and mitochondrial CRAT cDNAs and the location of an intron where the sequences diverge.

Sequence

Synthetic peptide located within the following region: MKASSRFKAHQDALPRLPVPPLQQSLDHYLKALQPIVSEEEWAHTKQLVD

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meghan E Ramsey et al.
Journal of bacteriology, 196(16), 2954-2968 (2014-06-11)
Neisseria gonorrhoeae uses a type IV secretion system (T4SS) to secrete chromosomal DNA into the medium, and this DNA is effective in transforming other gonococci via natural transformation. In addition, the T4SS is important in the initial stages of biofilm
Jee-Bum Lee et al.
Investigative ophthalmology & visual science, 55(7), 4119-4127 (2014-06-14)
To investigate the effects of visible light on human corneal epithelial cells and the impact of natural antioxidants on oxidative stress produced by overexposure to light. Light-emitting diodes with various wavelengths (410-830 nm) were used to irradiate human corneal epithelial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service