Skip to Content
Merck
All Photos(1)

Key Documents

93712

Sigma-Aldrich

Esterase Isoenzyme 3 porcine liver, recombinant

recombinant, expressed in E. coli, ≥0.8 U/mg

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
EC Number:
UNSPSC Code:
12352204
NACRES:
NA.54

recombinant

expressed in E. coli

form

crystalline
crystals
powder or flakes

specific activity

≥0.8 U/mg

storage temp.

−20°C

Application

Esterase Isoenzymes may be used to identify the monocytic element in normal and leukemic cells. They may be used for the biochemical characterization of different hematopoietic cell lineages and stages of differentiation when studying leukemias and lymphomas.

Biochem/physiol Actions

An esterase is a hydrolase that splits esters into acids and alcohols.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Unit Definition

1 U corresponds to the amount of enzyme which converts 1 μmol 4-nitrophenyl-L-acetate per minute at pH 7.5 and 30°C.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H G Drexler et al.
Cancer, 59(1), 77-82 (1987-01-01)
The expression of a particular alpha-naphthyl acetate esterase isoenzyme which is specific for monocytes was examined in a panel of cultured leukemia-lymphoma cell lines (n = 88), freshly obtained leukemia-lymphoma cells (n = 527), and in fresh (n = 10)
Esterase isoenzyme profiles of 255 leukemia-lymphoma cell lines from all hematopoietic cell lineages.
SM Gignac, ZB Hu, et al.
Leukemia Lymphoma, 22, 143-151 (1996)
B Sànchez-Nogué et al.
Environmental science and pollution research international, 20(5), 3480-3488 (2012-12-06)
The common sole, Solea solea (Linneus, 1758), and the Senegalese sole, Solea senegalensis (Kaup, 1858), are two important commercial species that coexist in the NW Mediterranean. In order to assess the species' ability to respond to chemical insults, a comparison
Marie C Fortin et al.
Drug metabolism and disposition: the biological fate of chemicals, 41(2), 326-331 (2012-12-12)
Studies on therapeutic drug disposition in humans have shown significant alterations as the result of pregnancy. However, it is not known whether pesticide metabolic capacity changes throughout pregnancy, which could affect exposure of the developing brain. We sought to determine
XieMei Tang et al.
Critical reviews in eukaryotic gene expression, 22(3), 179-187 (2012-11-13)
Tuberculosis remains one of the most prevalent and deadly infectious diseases, largely due to the emergence of multidrug-resistant and extensive drug-resistant Mycobacterium tuberculosis, especially the coinfection with HIV. Mycobacterium Ag85 complex (Ag85A, B, and C), with a carboxylesterase consensus sequence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service