95094
Heavy metal mix IV
(in 12% nitric acid), TraceCERT®, acc. to Nevada and Washington state regulation
About This Item
grade
certified reference material
TraceCERT®
Quality Level
product line
TraceCERT®
form
liquid
shelf life
limited shelf life, expiry date on the label
composition
As, 200 mg/L
Cd, 82 mg/L
Hg, 40 mg/L
Pb, 120 mg/L
concentration
(in 12% nitric acid)
technique(s)
ICP: suitable
gas chromatography (GC): suitable
application(s)
cannabis testing
format
multi-component solution
General description
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.
Application
Suitable for ICP-OES and ICP-MS
Preparation Note
Legal Information
Not finding the right product?
Try our Product Selector Tool.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B
Supplementary Hazards
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Ensure the safety of cannabis and hemp products by testing for heavy metal contamination using ICP-MS and state specific reference material mixes
A 4-step comprehensive ICP-MS workflow including sample homogenization, with standard addition calibration method, to determine heavy metals in Cannabis sativa.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service