Skip to Content
Merck
All Photos(1)

Key Documents

12356

Sigma-Aldrich

Benzoic acid

tested according to Ph. Eur.

Synonym(s):

Acidum benzoicum

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COOH
CAS Number:
Molecular Weight:
122.12
Beilstein:
636131
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Agency

USP/NF
tested according to Ph. Eur.

vapor density

4.21 (vs air)

vapor pressure

10 mmHg ( 132 °C)

Assay

99.9%

form

crystalline

autoignition temp.

1061 °F

bp

249 °C (lit.)

mp

121-125 °C (lit.)

solubility

water: soluble (2.9 g/l at 25 °C)

SMILES string

OC(=O)c1ccccc1

InChI

1S/C7H6O2/c8-7(9)6-4-2-1-3-5-6/h1-5H,(H,8,9)

InChI key

WPYMKLBDIGXBTP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzoic acid is an aromatic monocarboxylic acid. It occurs in the form of colorless leaflets or needles. It reacts with hydrogenating reagents to afford hexahydrobenzoic acid. On decomposition (by heating) in the presence of lime or alkali, it affords benzene and carbon dioxide. It can be synthesized by the cobalt or manganese catalyzed atmospheric oxidation of toluene.

Application

Benzoic acid may be employed as a standard in quantitative and calorimetric studies. It may be employed as an intermediate in the synthesis of the following:
  • paints
  • pigments
  • varnish
  • wetting agents
  • aroma compounds
  • benzoyl chloride
  • benzotrichloride
It was used to investigate the mechanism of complex addition reaction of hydroxyl radicals with various aromatic compounds.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hydroxylation by electrochemically generated OH. radicals. Mono-and polyhydroxylation of benzoic acid: products and isomer distribution.
Oturan MA and Pinson J.
The Journal of Physical Chemistry, 99(38), 13948-13954 (1995)
Eagleson M.
Concise Encyclopedia Chemistry, 122-122 (1994)
S Nacht et al.
Journal of the American Academy of Dermatology, 4(1), 31-37 (1981-01-01)
The transepidermal penetration and metabolic disposition of 14C-benzoyl peroxide were assessed in vitro (excised human skin) and in vivo (rhesus monkey). In vitro, the benzoyl peroxide penetrated into the skin, through the stratum corneum or the follicular openings, or both
G C Tremblay et al.
Pharmacology & therapeutics, 60(1), 63-90 (1993-10-01)
Detoxification of sodium benzoate by elimination as a conjugate with glycine, a nonessential amino acid, provides a pathway for the disposal of waste nitrogen. Since 1979, sodium benzoate has been widely used in the therapeutic regimen to combat ammonia toxicity
Renbo Wei et al.
Macromolecular rapid communications, 34(4), 330-334 (2013-01-03)
A new approach is developed to fabricate highly oriented mono-domain LCE nano/microstructures through micro-molding in capillaries. Gratings and microwires as two typical examples are fabricated and characterized by polarizing optical microscopy, optical microscopy, and scanning electron microscopy. The gratings with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service