SCC197
NP69 Human Nasopharyngeal Epithelial Cell Line
The NP69 cell line is useful for studies of the mechanisms of viral-associated tumorigenesis
Synonym(s):
NP69SV40T, NP-69
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
NACRES:
NA.45
Recommended Products
technique(s)
cell culture | mammalian: suitable
input
epithelial cells
General description
The NP69 immortalized human nasopharyngeal epithelial cell line retains many of the characteristics of normal nasopharyngeal cells, including keratin profiles and response to TGF-beta, while possessing many of the genetic signatures observed in NPC (2). NP69 cells are non-tumorigenic and demonstrate anchorage-dependent growth (2). The NP69 cell line is particularly useful for studies of the mechanisms of viral-associated tumorigenesis. Source: NP69 cells were derived from passage 1 cultures of biopsy-isolated primary nasopharyngeal epithelial cells transfected with the PX8 plasmid containing the simian virus 40 large T antigen (1).
Nasopharyngeal carcinoma (NPC), the most common cancer arising in the nasopharyngeal region, arises from epithelial cells and is especially prevalent in areas of southeast Asia and as a childhood cancer in some African populations (1). NPC ontology and tumorigenesis is closely associated with viral factors, especially Epstein-Barr virus (EBV) and human papillomavirus (HPV) (1).
References: 1. Lancet 2016; 387(10022): 1012-1024. 2. Biochim Biophys Acta 2002; 1590(1-3): 150-158.
Nasopharyngeal carcinoma (NPC), the most common cancer arising in the nasopharyngeal region, arises from epithelial cells and is especially prevalent in areas of southeast Asia and as a childhood cancer in some African populations (1). NPC ontology and tumorigenesis is closely associated with viral factors, especially Epstein-Barr virus (EBV) and human papillomavirus (HPV) (1).
References: 1. Lancet 2016; 387(10022): 1012-1024. 2. Biochim Biophys Acta 2002; 1590(1-3): 150-158.
Application
The NP69 cell line is useful for studies of the mechanisms of viral-associated tumorigenesis
This product is sold solely for research use per the terms of the “Restricted Use Agreement” which govern its use as detailed in the product Data Sheet or Certificate of Analysis. For information regarding any other use, please contact licensing@emdmillipore.com.
This product is sold solely for research use per the terms of the “Restricted Use Agreement” which govern its use as detailed in the product Data Sheet or Certificate of Analysis. For information regarding any other use, please contact licensing@emdmillipore.com.
Packaging
≥1X10⁶ cells/vial
Quality
- Each vial contains ≥ 1X10⁶ viable cells.
- Cells are tested negative for infectious diseases by a Human Essential CLEAR panel by Charles River Animal Diagnostic Services.
- Cells are verified to be of human origin and negative for inter-species contamination from rat, mouse, chinese hamster, Golden Syrian hamster, and non-human primate (NHP) as assessed by a Contamination CLEAR panel by Charles River Animal Diagnostic Services.
- Cells are negative for mycoplasma contamination.
- Each lot of cells is genotyped by STR analysis to verify the unique identity of the cell line.
Storage and Stability
Store in liquid nitrogen. The cells can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality.
Disclaimer
RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges Merck to request certain information about you and the establishment where the GMOs are being handled. Clickhere for Enduser Declaration (EUD) Form.
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service