Skip to Content
Merck
All Photos(1)

Key Documents

MAB10216

Sigma-Aldrich

Anti-Flavivirus Group Antigen Antibody, clone D1-4G2-4-15

ascites fluid, clone D1-4G2-4-15, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

mouse

antibody form

ascites fluid

antibody product type

primary antibodies

clone

D1-4G2-4-15, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable

isotype

IgG1
IgG2a

shipped in

dry ice

General description

The viruses of the family Flaviviridae are important arthropod-borne viruses in both human and veterinary medicine. They are transmitted by mosquito and ticks and usually are maintained in a transmission cycle in nature. They produce a broad spectrum of clinical responses in humans ranging from asymptomatic infection to fulminant encephalitis or haemorrhagic fever. Nearly 60 flaviviruses are known to exist but many are yet to be shown to cause disease in humans.

Specificity

Recognises flavivirus group specific antigens (Dengue virus, West Nile Virus, Japanese Encephalitis, Yellow Fever Virus, Zika virus etc). It binds to the fusion loop at the extremity of domain II of protein E.

Immunogen

Dengue virus type 2 antigens

Application

Anti-Flavivirus Group Antigen Antibody, clone D1-4G2-4-15 detects level of Flavivirus Group Antigen & has been published & validated for use in IF.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qian Chen et al.
Nature communications, 11(1), 2967-2967 (2020-06-13)
The recent outbreak of Zika virus (ZIKV) was associated with birth defects and pregnancy loss when maternal infection occurs in early pregnancy, but specific mechanisms driving placental insufficiency and subsequent ZIKV-mediated pathogenesis remain unclear. Here we show, using large scale
V Haridas et al.
PLoS neglected tropical diseases, 7(1), e2005-e2005 (2013-01-26)
Japanese encephalitis virus (JEV) is a major cause of viral encephalitis in South and South-East Asia. Lack of antivirals and non-availability of affordable vaccines in these endemic areas are a major setback in combating JEV and other closely related viruses
Sofie Jacobs et al.
International journal of molecular sciences, 20(9) (2019-04-30)
In humans, Zika virus and viral RNA have been detected in semen up to 2.2 months and 6 months post infection (pi), respectively. Although the contribution of sexual transmission to the spread of ZIKV is too low to sustain an
Andreas S Puschnik et al.
Cell reports, 21(11), 3032-3039 (2017-12-16)
The mosquito-borne flaviviruses include important human pathogens such as dengue, Zika, West Nile, and yellow fever viruses, which pose a serious threat for global health. Recent genetic screens identified endoplasmic reticulum (ER)-membrane multiprotein complexes, including the oligosaccharyltransferase (OST) complex, as
José A Aguilar-Briseño et al.
Nature communications, 11(1), 3177-3177 (2020-06-25)
Vascular permeability and plasma leakage are immune-pathologies of severe dengue virus (DENV) infection, but the mechanisms underlying the exacerbated inflammation during DENV pathogenesis are unclear. Here, we demonstrate that TLR2, together with its co-receptors CD14 and TLR6, is an innate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service