Skip to Content
Merck
All Photos(1)

Key Documents

IPAKMETA1

Milli-Q

IPAK Meta® Polishing Cartridge

Designed for use with Milli-Q® IQ 7000 and Milli-Q® EQ 7000/08/16 ultrapure water systems

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104212
eCl@ss:
33050190

Quality Level

packaging

pkg of 1 unit

compatibility

for use with Milli-Q® EQ 7000
for use with Milli-Q® EQ 7008
for use with Milli-Q® EQ 7016
for use with Milli-Q® IQ 7000

shipped in

ambient

General description

IPAK Meta® water polishing cartridge is designed to work as a pair, with the IPAK Quanta® polishing cartridge, for the production of ultrapure water in Milli-Q® IQ 7000 and Milli-Q® EQ 7000/08/16 water purification systems. These systems provide superior-quality ultrapure water for sensitive research applications.
The cartridges use a combination of Jetpore® mixed-bed ion-exchange resin and innovative IQnano ion-exchange media to remove trace ions from pure water, providing exceptional performance.

Features and Benefits

  • A full report is available for the IPAK Meta® cartridges. It provides details on the technologies used, their performance during testing by our R&D department, as well as information concerning our quality and manufacturing requirements.
  • e-Sure tag for RFID connection with POD enables full traceability (data management) and automatic consumable status monitoring on the POD′s touchscreen interface.
  • 78% of the parts suppliers are ISO 14001 environmental management system-certified.

Legal Information

IPAK META is a registered trademark of Merck KGaA, Darmstadt, Germany
IPAK QUANTA is a registered trademark of Merck KGaA, Darmstadt, Germany
IQNANO is a trademark of Merck KGaA, Darmstadt, Germany
JETPORE is a registered trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service