Skip to Content
Merck
All Photos(2)

Key Documents

HTS123M

Sigma-Aldrich

ChemiScreen Recombinant Human BB1 Bombesin/Neuromedin B Receptor Membrane Preparation

Human BB1 GPCR membrane preparation for Radioligand binding Assays & GTPγS binding.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32161000
NACRES:
NA.84

biological source

human

Quality Level

recombinant

expressed in Chem-1 cells

manufacturer/tradename

ChemiScreen
Chemicon®

technique(s)

ligand binding assay: suitable (GTPγS)
radioligand binding assay (RLBA): suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Gene Information

human ... NMBR(4829)

General description

Bombesin, a bioactive peptide first identified in amphibian skin, is related to two mammalian peptides, gastrin-releasing peptide (GRP) and neuromedin B (NMB). A family of 3 GPCRs, including NMB-R (BB1), GRP-R (BB2), and BRS-3 (BB3), mediate the biological effects of the peptides. The receptors differ in their affinities for the peptides; BB2 binds to GRP with 50-300-fold greater affinity than to NMB, whereas BB1 binds to NMB with 10-800-fold greater affinity than to GRP (Tokita et al., 2004). Binding of ligand to BB1 activates Gq to increase intracellular calcium concentrations. The CNS is a major site of NMB and BB1 expression, and BB1 appears to be involved in thermoregulation (Ohki-Hamazaki et al., 2005). Chemicon′s BB1 membrane preparations are crude membrane preparations made from our proprietary stable recombinant cell lines to ensure high-level of GPCR surface expression; thus, they are ideal HTS tools for screening of antagonists of BB1 interactions with neuromedin B. The membrane preparations exhibit a Kd of 0.35-0.43 nM for [125I]-Bombesin. With 5 µg/well BB1 Membrane Prep and 0.3 nM [125I]-bombesin, a greater than 40-fold signal-to-background ratio was obtained.
Full-length human NMBR cDNA encoding BB1

Application

Radioligand binding assay, and GTPγS binding.

Biochem/physiol Actions

GPCR Class: A
Protein Target: BB1
Target Sub-Family: Bombesin

Packaging

1 mL per vial

Quality

Signal:background and specific binding values obtained in a competition binding assay with varying amounts of BB1 membrane prep:
10 µg/well5 µg/well
Signal:Background 82.5 74.8
Specific Binding (cpm) 27377 22443


SPECIFICATIONS: 1 unit = 5 µg membrane preparation
Bmax: 3.1 pmol/mg
Kd: 0.39 nM

Specifications

Inucbation Conditions
Membranes are mixed with radioactive ligand and unlabeled competitor (see Figures 1 and 2 for concentrations tested) in binding buffer in a nonbinding 96-well plate, and incubated for 1-2 h. Prior to filtration, a GF/C 96-well filter plate is coated with 0.33% polyethyleneimine for 30 min, then washed with 50mM HEPES, pH 7.4, 0.5% BSA. Binding reaction is transferred to the filter plate, and washed 3 times (1 mL per well per wash) with Wash Buffer. The plate is dried and counted.

Binding buffer: 50 mM Hepes, pH 7.4, 5 mM MgCl2, 1 mM CaCl2, 0.2% BSA, filtered and stored at 4°C
Radioligand: [125I] bombesin (Perkin Elmer # NEX258)
Wash Buffer: 50 mM Hepes, pH 7.4, 500mM NaCl , 0.1% BSA, filtered and stored at 4°C.

Physical form

One package contains enough membranes for at least 200 assays (units), where an unit is the amount of membrane that will yield greater than 40-fold signal:background with 125I-labeled bombesin at 0.3 nM.

Liquid in packaging buffer: 50 mM Tris pH 7.4, 10% glycerol and 1% BSA with no preservatives.
Packaging method: Membranes protein were adjusted to the indicated concentration in packaging buffer, rapidly frozen, and stored at -80°C.

Storage and Stability

Maintain frozen at -70°C for up to 2 years. Do not freeze and thaw.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular basis of the selectivity of gastrin-releasing peptide receptor for gastrin-releasing peptide
Tokita, Kenji, et al
Molecular Pharmacology, 61, 1435-1443 (2002)
Development and function of bombesin-like peptides and their receptors
Ohki-Hamazaki, Hiroko, et al
International Journal of Developmental Biology, 49, 293-300 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service