Skip to Content
Merck
All Photos(1)

Key Documents

AG235

Sigma-Aldrich

Dopamine D4 Peptide

control for AB1787P, liquid, mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.32

Pricing and availability is not currently available.

Product Name

Dopamine D4, Mouse, Ms, Control Peptide for AB1787P,

biological source

mouse

Quality Level

form

liquid

manufacturer/tradename

Chemicon®

concentration

1.0 mg/mL

technique(s)

cell based assay: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
GF164SRP3007GF315
impurities

<0.1 μg/mg protein endotoxin (<1EU/μg)

impurities

<0.1 μg/mg protein endotoxin (<1EU/μg)

impurities

<0.1 EU/μg endotoxin, tested

impurities

<0.1 μg/mg protein endotoxin (<1EU/μg)

shipped in

dry ice

shipped in

dry ice

shipped in

wet ice

shipped in

dry ice

UniProt accession no.

O15123

UniProt accession no.

Q15389

UniProt accession no.

Q15389

UniProt accession no.

P15692

Quality Level

100

Quality Level

100

Quality Level

-

Quality Level

100

General description

10 amino acid peptide sequence from the mouse D4 receptor within the 3rd extracellular domain.

Application

For use in blocking the reactivity of AB1787P.

Optimal working dilution must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors

Physical form

Liquid in PBS, pH 7.4 containing 0.02% sodium merthiolate.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2


  • Certificates of Analysis (COA)

    Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    C D Livingstone et al.
    The Biochemical journal, 287 ( Pt 1), 277-282 (1992-10-01)
    Three-dimensional computer models of the rat D2, D3 and D4 dopamine receptor subtypes have been constructed based on the diffraction co-ordinates for bacteriorhodopsin, another membrane-bound protein containing seven transmembrane domains presumed to be arranged in a similar spatial orientation. Models
    H H Van Tol et al.
    Nature, 358(6382), 149-152 (1992-07-09)
    The dopamine D4 receptor structurally and pharmacologically resembles the dopamine D2 and D3 receptors. Clozapine, an atypical antipsychotic that is relatively free of the adverse effects of drug-induced parkinsonism and tardive dyskinesia, binds to the D4 receptor with an affinity
    H H Van Tol et al.
    Nature, 350(6319), 610-614 (1991-04-18)
    Dopamine receptors belong to the family of G protein-coupled receptors. On the basis of the homology between these receptors, three different dopamine receptors (D1, D2, D3) have been cloned. Dopamine receptors are primary targets for drugs used in the treatment

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service