Skip to Content
Merck
All Photos(1)

Key Documents

5.04042

Millipore

Doxorubicin Hydrochloride

InSolution, ≥98%

Synonym(s):

InSolution Doxorubicin, Hydrochloride, Adriamycin, HCl, 14-Hydroxydaunomycin, HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H29NO11 · xHCl
CAS Number:
Molecular Weight:
543.52 (free base basis)
MDL number:
UNSPSC Code:
12352101

Quality Level

Assay

≥98% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
protect from light

storage temp.

−70°C

InChI

1S/C27H29NO11.ClH/c1-10-22(31)13(28)6-17(38-10)39-15-8-27(36,16(30)9-29)7-12-19(15)26(35)21-20(24(12)33)23(32)11-4-3-5-14(37-2)18(11)25(21)34;/h3-5,10,13,15,17,22,29,31,33,35-36H,6-9,28H2,1-2H3;1H/t10?,13?,15-,17?,22?,27-;/m0./s1

InChI key

MWWSFMDVAYGXBV-FGBJBKNOSA-N

General description

A 10 mM (5 mg/862 µL) sterile-filtered solution of Doxorubicin, HCl in H₂O.
An anti-tumor antibiotic and a highly effective myotoxin that inhibits topoisomerase II (IC50 = 100 nM). Binds to nucleic acids, presumably by specific intercalation into the DNA double helix, inhibiting nucleic acid synthesis. Cytotoxicity seems to be due to its ability to intercalate with DNA, interact with plasma membranes, and take part in oxidation-reduction reactions. Induces apoptosis in rhabdomyosarcoma cell lines.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Physical form

A 10 mM (5 mg/862 µL) sterile-filtered solution of Doxorubicin, HCl (Cat. No. 324380) in H₂O.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C or -70°C). Aliquots are stable for up to 3 months at -20°C or for up to 6 months at -70°C.

Other Notes

A′Hern, R.P., and Gore, M.E. 1995.J. Clin. Oncol.13, 726.
Nooter, K., et al. 1995. Br. J. Cancer71, 556.
Noviello, E., et al. 1994. Mutat. Res.. 311, 21.
Anderson, R.D., et al. 1993. Mutat. Res.294, 215.
Hershko, C., et al. 1993. J. Lab. Clin. Med.122, 245.
Jongmans, W., et al. 1993. Mutat. Res.294, 207.
O′Shaughnessy, J.A., and Cowan, K.H. 1993. J. Am. Med. Assoc.270, 2089.
Theyer, G., et al. 1993. J. Urol.150, 1544.
Tritton, T.R., and Yee, G. 1982. Science217, 248.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service