Skip to Content
Merck
All Photos(1)

Key Documents

1.12167

Sigma-Aldrich

Gadolinium(III) oxide

99+

Synonym(s):

Gadolinium(III) oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Gd2O3
CAS Number:
Molecular Weight:
362.50
MDL number:
UNSPSC Code:
12352303
EC Index Number:
235-060-9

Quality Level

potency

>5000 mg/kg LD50, oral (Rat)

mp

2330 °C

density

7.4 g/cm3 at 20 °C

storage temp.

no temp limit

InChI

1S/2Gd.3O/q2*+3;3*-2

InChI key

CMIHHWBVHJVIGI-UHFFFAOYSA-N

Analysis Note

Assay (complexometric, calc. on the ignited basis): ≥ 99.0 %
Identity (ICP-OES): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service