Skip to Content
Merck
All Photos(3)

Key Documents

07-1224

Sigma-Aldrich

Anti-PP6C Antibody

from rabbit

Synonym(s):

H.sapiens mRNA for protein phosphatase 6, Serine/threonine-protein phosphatase 6, Protein phosphatase 6, catalytic subunit, Serine/threonine protein phosphatase catalytic subunit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
IHC
IP
WB
species reactivity:
mouse, rat, bovine, human
technique(s):
immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
western blot: suitable
citations:
5

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, rat, bovine, human

technique(s)

immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PPP6C(5537)

General description

This gene encodes the catalytic subunit of protein phosphatase, a component of a signaling pathway regulating cell cycle progression.

Specificity

Recognizes the catalytic subunit of Protein Phosphatase 6, MW ~33kDa.

Immunogen

The immunogen used to the purified peptide conjugated to KLH corresponding to the sequence NH2-Cys-Ala-Val-Pro-Asp-Ser-Glu-Arg-Val-Ile- Pro-Pro-Arg-Thr-Thr-Thr-Pro-Tyr-COOH.

Application

Immunoprecipitation: A previous lot of this antibody was used in immunoprecipitation at a dilution of 10-20 µg/mL.
Immunohistochemistry(paraffin):
Optimal Staining of PP6 Polyclonal Antibody:
Human Testis
PP6 staining of round spermatids in normal human
testis, tissue pretreated with citrate buffer, pH 6.0.
Antibody was diluted 1:100, using IHC-Select
detection with HRP-DAB.
Detect PP6C using this Anti-PP6C Antibody validated for use in WB, IP, IH(P).
Research Category
Signaling
Research Sub Category
Kinases & Phosphatases

Quality

Routinely evaluated by Western Blot on Jurkat lysates.

Western Blot Analysis: 1:500 dilution of this lot detected PP6C on 10 μg of Jurkat lysates.

Target description

Recognizes the catalytic subunit of Protein Phosphatase 6 (PP6), MW ~33kDa

Linkage

Replaces: AB4091

Physical form

Format: Purified
Purified rabbit polyclonal IgG in buffer containing PBS with 0.08% sodium azide.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 1 year from date of receipt.
Handling Recommendations: Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Jurkat Cell Lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiangli Cui et al.
Carcinogenesis, 31(10), 1734-1741 (2010-08-24)
We have recently shown that American ginseng (AG) prevents and treats mouse colitis. Because both mice and humans with chronic colitis have a high colon cancer risk, we tested the hypothesis that AG can be used to prevent colitis-driven colon
Jordan Wengrod et al.
Science signaling, 8(367), ra27-ra27 (2015-03-12)
Amino acid deprivation promotes the inhibition of the kinase complex mTORC1 (mammalian target of rapamycin complex 1) and activation of the kinase GCN2 (general control nonrepressed 2). Signaling pathways downstream of both kinases have been thought to independently induce autophagy.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service