Skip to Content
Merck
All Photos(1)

Key Documents

WHA10347009

Whatman® Grade 8 ruled circles

size 90 mm

Synonym(s):

Z696269, filter paper, paper filter, whatman filter

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104900
NACRES:
NB.24

material

cellulose

feature

Grade 8

packaging

pkg of 100 ea

manufacturer/tradename

Whatman 10347009
Whatman Article No. 28414641 (US reference)

size

90 mm

basis weight

65

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Filter papre for routine investigations of foreign substances in a variety of sample types.

Features and Benefits

  • White filter paper with printed green lines for optical assessment.

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Legal Information

Whatman is a registered trademark of Cytiva

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service