Skip to Content
Merck
All Photos(2)

Key Documents

GF57977971

Silver

tube, 200mm, outside diameter 1.75mm, inside diameter 1.55mm, wall thickness 0.1mm, as drawn, 99.95+%

Synonym(s):

Silver, AG007150

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ag
CAS Number:
Molecular Weight:
107.87
MDL number:
UNSPSC Code:
12141740
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99.95%

form

tubes

manufacturer/tradename

Goodfellow 579-779-71

resistivity

1.59 μΩ-cm, 20°C

L × wall thickness

200 mm × 0.1 mm

O.D. × I.D.

1.75 mm × 1.55 mm

bp

2212 °C (lit.)

mp

960 °C (lit.)

density

10.49 g/cm3 (lit.)

SMILES string

[Ag]

InChI

1S/Ag

InChI key

BQCADISMDOOEFD-UHFFFAOYSA-N

Related Categories

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pakvirun Thuesombat et al.
Ecotoxicology and environmental safety, 104, 302-309 (2014-04-15)
With the advances in nanotechnology, silver nanoparticles (AgNPs) have been applied in many industries, increasing their potential exposure level in the environment, yet their environmental safety remains poorly evaluated. The possible effects of different sized AgNPs (20, 30-60, 70-120 and
Sisi Liu et al.
Journal of nanoscience and nanotechnology, 14(6), 4608-4614 (2014-04-18)
Position-configurable, reproducible, vertically aligned nanosheets assemblies (ANAs) arrays are fabricated by polymer-templated electrodeposition method at room temperature. Here, nanoimprint lithography is utilized to fabricate polymer template on the fluorine-doped tin oxide substrate for the purpose of evenly tuning the location
Yu Sun et al.
Journal of nanoscience and nanotechnology, 14(6), 4481-4485 (2014-04-18)
The fluorescence enhancement effect of Rh6G molecules deposited on the silver film substrate decorated with nanohole arrays was investigated in this paper. The prepared substrate, decorated with nanohole arrays, was fabricated with the deposition of silver films onto the anodic
Hajo Haase et al.
Journal of biomedical nanotechnology, 10(6), 1146-1156 (2014-04-23)
Silver is commonly used as an antibacterial agent, e.g., in various medical applications, and the availability of silver nanoparticles (AgNP) has fueled this development. Their antibacterial properties are well defined, whereas there are concerns regarding unknown and potentially harmful effects
Swarup Roy et al.
Journal of nanoscience and nanotechnology, 14(7), 4899-4905 (2014-04-25)
Binding interaction of biologically synthesized silver nanoparticles with bovine serum albumin (BSA) has been investigated by UV-Vis and fluorescence spectroscopic techniques. UV-Vis analysis implies the formation of the ground state complex between BSA and silver nanoparticles. The analysis of fluorescence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service