BAF371220045
SP Bel-Art® Spinbar® magnetic stir bar
polygon white PTFE-coated, L × diam. 1.75 in. (45 mm) × 0.3125 in. (8 mm)
Synonym(s):
Spinbar® magnetic stir bar
All Photos(7)
About This Item
UNSPSC Code:
41122800
NACRES:
NB.51
Recommended Products
material
polygon white PTFE-coated
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F37122-0045
L × diam.
1.75 in. (45 mm) × 0.3125 in. (8 mm)
General description
- Spinbar® magnetic stir bar is used in conjunction with a magnetic stirring plate for uniform, consistent, and thorough mixing of chemical solutions.
- Alnico magnet encapsulated in durable and inert FDA grade polytetrafluoroethylene (PTFE) low friction coating.
- Multi-faceted-shaped bars provide a greater surface area and added turbulence for more vigorous, thorough mixing.
- Autoclave safe.
Leakproof, seamless, polished, and contains a permanent Alnico V magnet. Egg-shape bar is designed especially for use in round-bottom vessels.
These stirbars have polygon multifaceted surfaces that add turbulence relative to similar smooth size cylindrical bars.
Features and Benefits
- Center molded pivot ring minimizes the contact area between the bar and the vessel; reduces friction and chattering
- Marring of plastic containers is diminished
- Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE coating – Superior Chemical Compatibility
- Manufactured in a registered ISO 9001:2015 facility, as verified by SGS certification
- Autoclave safe
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Spinbar is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)