Skip to Content
Merck
All Photos(6)

Key Documents

806641

Sigma-Aldrich

Graphene nanoplatelets

powder, oxidized

Synonym(s):

GNPs, Graphenit-OX

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
Molecular Weight:
12.01
UNSPSC Code:
12352103
NACRES:
NA.23

description

Dispersibility: Water, THF, DMF

form

powder

composition

Carbon, >95 wt. %
Oxygen, >1 wt. %

surface area

110 m2/g , BET

conductivity

1.5 E+05 S/m (pressed pellets)

bulk density

0.2 g/mL

Looking for similar products? Visit Product Comparison Guide

General description

A low dimensional carbon nanoplatelet with a lateral dimension of about 2-3μ and a few layer thick. It is slightly oxidized (content of oxygen around 2%) yielding to a superior dispersion behavior in different solvent/matrix. It has been used with success as anti-corrosion additive in powder coating formulation among other properties.

Application

  • Graphene (nano)composite materials
  • Conductive coatings
  • Anti-corrosion coatings
  • Conductive Inks
  • Energy Storage
  • Electrode materials

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Facile fabrication of highly flexible graphene paper for high-performance flexible lithium ion battery anode
Kim et al.
Royal Society of Chemistry Advances, 5(5), 3299-3305 (2015)
New Approach to the Synthesis of Exfoliated Polymer/Graphite Nanocomposites by Miniemulsion Polymerization Using Functionalized Graphene
Han et al
Macromolecules, 44(21), 8504-8515 (2011)
Yan Geng et al.
Journal of colloid and interface science, 336(2), 592-598 (2009-05-06)
A novel route is proposed to produce graphite nanoplatelets (GNPs) and graphene sheets. The natural graphite flakes were directly exfoliated by ultrasonication in formic acid. A stable graphene aqueous dispersion was obtained using the as-produced GNPs after two processing steps:
Ladislav Kavan et al.
ACS nano, 5(1), 165-172 (2010-12-04)
Commercial graphene nanoplatelets in the form of optically transparent thin films on F-doped SnO(2) (FTO) exhibited high electrocatalytic activity toward I(3)(-)/I(-) redox couple, particularly in electrolyte based on ionic liquid (Z952). The charge-transfer resistance, R(CT), was smaller by a factor

Articles

Since its discovery little more than a decade ago,1 the two-dimensional (2D) allotrope of carbon—graphene—has been the subject of intense multidisciplinary research efforts.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service