Skip to Content
Merck
All Photos(4)

Key Documents

805866

Sigma-Aldrich

i-Butylammonium iodide

greener alternative

Synonym(s):

2-Methylpropan-1-aminium iodide, i-Butylamine hydriodide, Greatcell Solar®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H12IN
Molecular Weight:
201.05
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

168.2 °C

greener alternative category

SMILES string

CC(C)CN.I

InChI

1S/C4H11N.HI/c1-4(2)3-5;/h4H,3,5H2,1-2H3;1H

InChI key

FCTHQYIDLRRROX-UHFFFAOYSA-N

General description

i-Butlyammonium iodide (iso-BAI) is an ammonium salt that facilitates the formation of highly stable perovskites with good performance. These perovskite materials act as moisture resistant light absorbers. The butylamine is used as a spacing ammonium cation that supports the insulation of 2D perovskites.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more details.

Application

Iso-BAI in combination with formamidinium iodide (FAI) forms a mixture of bulky organic ammonium iodide, which is used in the fabrication of perovskite based solar cells with a power conversion efficiency of > 21%.
The iodide and bromide based alkylated halides find applications as precursors for fabrication of perovskites for photovoltaic applications.

Legal Information

Product of Greatcell Solar®
Greatcell Solar is a registered trademark of Greatcell Solar

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hydrogen bond enables highly efficient and stable two-dimensional perovskite solar cells based on 4-pyridine-ethylamine
Li J, et al.
Organic Electronics, 67(24), 122-127 (2019)
Recent progress in 2D/quasi-2D layered metal halide perovskites for solar cells
Yan J, et al.
Journal of Material Chemistry A, 6(24), 11063-11077 (2018)
Nam Joong Jeon et al.
Nature, 517(7535), 476-480 (2015-01-07)
Of the many materials and methodologies aimed at producing low-cost, efficient photovoltaic cells, inorganic-organic lead halide perovskite materials appear particularly promising for next-generation solar devices owing to their high power conversion efficiency. The highest efficiencies reported for perovskite solar cells
Wei Zhang et al.
Nano letters, 15(3), 1698-1702 (2015-02-05)
The performance of perovskite solar cells has been progressing over the past few years and efficiency is likely to continue to increase. However, a negative aspect for the integration of perovskite solar cells in the built environment is that the
Zhi-Kuang Tan et al.
Nature nanotechnology, 9(9), 687-692 (2014-08-05)
Solid-state light-emitting devices based on direct-bandgap semiconductors have, over the past two decades, been utilized as energy-efficient sources of lighting. However, fabrication of these devices typically relies on expensive high-temperature and high-vacuum processes, rendering them uneconomical for use in large-area

Articles

Next generation solar cells have the potential to achieve conversion efficiencies beyond the Shockley-Queisser (S-Q) limit while also significantly lowering production costs.

Dr. Perini and Professor Correa-Baena discuss the latest research and effort to obtain higher performance and stability of perovskite materials.

For several decades, the need for an environmentally sustainable and commercially viable source of energy has driven extensive research aimed at achieving high efficiency power generation systems that can be manufactured at low cost.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service