Skip to Content
Merck
All Photos(2)

Key Documents

802158

Sigma-Aldrich

Polycaprolactone dimethacrylate

average Mn 3,000

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H10O2)nC12H18O5
UNSPSC Code:
12162002

form

powder

mol wt

average Mn 3,000

mp

38-43 °C

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Polycaprolactone is a biodegradable polymer commonly used in biomedical applications. A light-activated Polycaprolactone dimethacrylate gel was used for the sustained delivery of an anti-VEGF antibody.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Puneet Tyagi et al.
Molecular pharmaceutics, 10(8), 2858-2867 (2013-06-06)
A light-activated polycaprolactone dimethacrylate (PCM) and hydroxyethyl methacrylate (HEMA) based gel network was developed to sustain the release of stable, active bevacizumab (an anti-VEGF antibody used to treat choroidal neovascularization) and used to assess sustained ex vivo delivery in rabbit

Articles

We will explore the technological advances that have contributed toward the progress of 3DP of tissue engineering scaffolds, current materials used to create 3DP scaffolds, and the challenges that remain.

Innovations in polymer technology have had a significant impact on the advancement of novel drug delivery systems.

In the past two decades, tissue engineering and regenerative medicine have become important interdisciplinary fields that span biology, chemistry, engineering, and medicine.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service