Skip to Content
Merck
All Photos(1)

Key Documents

777986

Sigma-Aldrich

CdTe core-type quantum dots

COOH functionalized, fluorescence λem 510 nm, powder

Synonym(s):

Fluorescent nanocrystals, QDs, artificial atoms

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CdTe
EC Number:
UNSPSC Code:
26111700

form

powder

fluorescence

λem 510 nm
, quantum yield 37%

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Cadmium Telluride Quantum Dots (CdTe QDs) are available in solid form. They are coated with carboxyl groups which makes them water soluble to make aqueous solutions. These carboxyl functionalized hydrophilic quantum dots can be further conjugated for use as labels. CdTe QDs can be used for optical and display applications; solar cells and biomedical applications.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R S Aga et al.
Nanotechnology, 20(46), 465204-465204 (2009-10-23)
A photosensitized high-surface area transparent electrode has been employed to increase the short circuit current of a photovoltaic device with a blend of poly(3-hexylthiophene) (P3HT) and (6,6)-phenyl C61 butyric acid methyl ester (PCBM) as the active layer. This is achieved
P Haro-González et al.
Small (Weinheim an der Bergstrasse, Germany), 8(17), 2652-2658 (2012-06-16)
The potential use of CdTe quantum dots as luminescence nano-probes for lifetime fluorescence nano-thermometry is demonstrated. The maximum thermal sensitivity achievable is strongly dependent on the quantum dot size. For the smallest sizes (close to 1 nm) the lifetime thermal
A Rameshkumar et al.
Colloids and surfaces. B, Biointerfaces, 101, 74-82 (2012-07-17)
This study was undertaken to evaluate the antioxidant potential of an aqueous extract from Merremia emarginata leaves because this plant has a very high flavonoid and phenol content. The in vitro antioxidant activity was measured by diphenyl-1-picrylhydrazyl (DPPH), 2,2'-azino-bis (3-ethylbenzthiazoline-6-sulphonic

Articles

Since the first report of the low-cost dye-sensitized solar cell (DSSC) in 1991 by Gratzel and his coworker,1 dye-sensitized solar cells (DSSC) has been regarded as one of the most promising photovoltaic technologies because of their transparent and colorful characteristics, as well as low cost.

Professor Sharma and colleagues review the synthesis and applications of this novel material. This includes a discussion of the unique properties of quantum dots and their suitability for solar cell applications, along with common synthesis techniques used to develop these materials.

Professor Xiaohu Gao (University of Washington, USA) provides a overview of recent quantum dot (QD) advancements and their potential for advancing bioassay and bioimaging technologies.

Studies on perovskite quantum dots are developing and progress has been made toward overcoming the challenges of QDs. Strategies to fabricate high-quality materials have enabled rapid light-emitting device development.

See All

Protocols

Selenium is an essential trace element. It is a necessary dietary constituent of at least 25 human selenoproteins and enzymes containing selenocysteine. Additionally, as selenium is a semiconductor and photoelectrically active, it has more advanced applications such as xerography and solar cell assembly.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service