Skip to Content
Merck
All Photos(1)

Key Documents

773395

Sigma-Aldrich

Poly(L-lactide), 2-bromoisobutyryl terminated

average Mn 5,500, PDI ≤1.1

Synonym(s):

ATRP Macromonomer, ATRP terminated PLLA, PLA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H(C3H4O2)nC6H10BrO3
UNSPSC Code:
12162002
NACRES:
NA.23

form

solid

optical activity

[α]22/D -133.0°, c = 0.1% in chloroform

mol wt

average Mn 5,500

PDI

≤1.1

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Macroinitiator for atom transfer radical polymerization (ATRP) to prepare block copolymers. Poly(L-lactide) is commonly used in drug delivery.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oh; Jung Kwon.
Soft Matter, 7, 5096-5096 (2011)

Articles

Local delivery of bioactive molecules using an implantable device can decrease the amount of drug dose required as well as non-target site toxicities compared to oral or systemic drug administration.

Microparticle drug delivery systems have been extensively researched and applied to a wide variety of pharmaceutical and medical applications due to a number of advantages including injectability, local applicability to target tissues and sites, and controlled drug delivery over a given time period.

Wide range of functional polymers for biomedical applications have been synthesized and structurally characterized. Several classes of polymers including biodegradable polymers, hydrophilic & amphiphilic polymers, and stimuli responsive polymers have been prepared using controlled and directed functionalization via "living" polymerization such as RAFT, ionic and ring opening polymerization. Selected polymers have been studied for their structure-properties relationship. "

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service