Skip to Content
Merck
All Photos(3)

Key Documents

752630

Sigma-Aldrich

Magnesium silicide

greener alternative

pieces, 99.7% trace metals basis

Synonym(s):

Dimagnesium silicide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Mg2Si
CAS Number:
Molecular Weight:
76.70
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

99.7% trace metals basis

form

pieces

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

particle size

3-14 mm

greener alternative category

SMILES string

[Mg]=[Si]=[Mg]

InChI

1S/2Mg.Si

InChI key

YTHCQFKNFVSQBC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Magnesium silicide (Mg2Si) can be used as a thermoelectric material for the fabrication of thermoelectric (TE) devices. It can also be used as a counter electrode for lithium-ion batteries. It can generally be synthesized as a nanoparticle that can be utilized as a deoxygenation agent.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Water-react 2

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sakamoto, T.; et al.
Journal of Electronic Materials, 39, 1708-1708 (2010)
Magnesium silicide as a negative electrode material for lithium-ion batteries
Reimer JA
Journal of Power Sources, 4819, 1-6 null
Etienne Savary et al.
Dalton transactions (Cambridge, England : 2003), 39(45), 11074-11080 (2010-10-23)
The ultra fast synthesis of nanocrystalline Mg(2)Si was carried out using microwave radiation. The elemental precursors were first milled together under dry conditions to get fine particles. The resulting mixture of powders of Mg and Si was cold pressed before
Ihou-Mouko, H.; Mercier, C.; Tobola, J.; Pont, G.; Scherrer, H.
J. Alloy Compounds, 509, 6503-6503 (2011)

Articles

The price of tellurium, a key component in many thermoelectric materials, has risen in recent years, leading to the search for more cost-effective substitutes. This article presents silicide materials as a cheaper potential alternative.

Higher transition metal silicides are very well-suited for anisotropic thermoelectric conversion. Essential anisotropy of the Seebeck coefficient, together with good mechanical properties, allows production of reliable anisotropic thermoelectric converters.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service