Skip to Content
Merck
All Photos(1)

Key Documents

646202

Sigma-Aldrich

Thymidine-13C10,15N2 5′-triphosphate disodium salt solution

100 mM (in 5mM Tris HCl / H2O), ≥98 atom %, ≥95% (CP)

Synonym(s):

Deoxythymidine 5′-triphosphate-13C10,15N2 disodium salt solution, TTP-13C10,15N2 disodium salt solution, dTTP-13C10,15N2 disodium salt solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C10H1515N2Na2O14P3
Molecular Weight:
538.05
UNSPSC Code:
12352200
PubChem Substance ID:

isotopic purity

 15N
≥98 atom %

Quality Level

Assay

≥95% (CP)

concentration

100 mM (in 5mM Tris HCl / H2O)

technique(s)

bio NMR: suitable

mass shift

M+12

shipped in

dry ice

storage temp.

−20°C

SMILES string

[Na+].[13CH3][13C]1=[13CH][15N]([13C@H]2[13CH2][13C@H](O)[13C@@H]([13CH2]OP(O)(=O)OP(O)(=O)OP(O)(O)=O)O2)[13C](=O)[15NH][13C]1=O

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Max A Horlbeck et al.
Cell, 174(4), 953-967 (2018-07-24)
Seminal yeast studies have established the value of comprehensively mapping genetic interactions (GIs) for inferring gene function. Efforts in human cells using focused gene sets underscore the utility of this approach, but the feasibility of generating large-scale, diverse human GI

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service