Skip to Content
Merck
All Photos(2)

Key Documents

545015

Sigma-Aldrich

Silver nitrite

99.98% trace metals basis

Synonym(s):

Silver nitrite (AgNO), Silver(I) nitrite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
AgNO2
CAS Number:
Molecular Weight:
153.87
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.98% trace metals basis
form:
powder

Assay

99.98% trace metals basis

form

powder

impurities

≤250.0 ppm Trace Metal Analysis

mp

140 °C (lit.)

density

4.453 g/mL at 25 °C (lit.)

SMILES string

[Ag+].[O-]N=O

InChI

1S/Ag.HNO2/c;2-1-3/h;(H,2,3)/q+1;/p-1

InChI key

KKKDGYXNGYJJRX-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Silver nanomaterials have unique physical, chemical, and optical properties that are currently being leveraged for a wide variety of biological applications.

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service