Skip to Content
Merck
All Photos(3)

Key Documents

HPA007729

Sigma-Aldrich

Anti-LTA antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-LTα, Anti-Lymphotoxin-α, Anti-Tumor necrosis factor ligand superfamily member 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

HSTLKPAAHLIGDPSKQNSLLWRANTDRAFLQDGFSLSNNSLLVPTSGIYFVYSQVVFSGKAYSPKATSSPLYLAHEVQLFSSQYPFHVPLLSSQKMVYPGLQEPWLHSMYHGAAFQLTQGDQLSTHTDGIPHLVLSP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LTA(4049)

General description

LTA (Lymphotoxin-alpha) gene encodes a member of the TNF (tumor necrosis factor) superfamily. It is also called as TNFβ and is secreted as a homotrimer by activated lymphocytes.

Immunogen

LTA protein Lymphotoxin α recombinant protein epitope signature tag (PrEST)

Application

Anti-LTA antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

LTA (Lymphotoxin-α) forms a heterotrimer with lymphotoxin-β (LT-β), which anchors it to the cell surface. It binds to either TNF-R55 or TNF-R75 receptors (TNF receptors) and mediates inflammatory, immunostimulatory, and antiviral responses or host defense responses. When complexed with LTβ, it does not bind to the TNF receptors. The LTα/β complex binds to a unique receptor called the LTβ-R. It is also involved in the development or organization of peripheral lymphoid organs. Defects in this gene are associated with susceptibility to leprosy type 4, myocardial infarction, non-Hodgkin′s lymphoma, and psoriatic arthritis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70026

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Na Li et al.
PloS one, 9(3), e92272-e92272 (2014-03-20)
The lymphotoxin-α (LTA), as one of the mediators of inflammation, may play an important role in the pathogenesis of myocardial infarction (MI). Genetic association studies (GAS) that have investigated the association between three common polymorphisms (A252G, G10A and C804A) of
K A Elisa Kallio et al.
Infection and immunity, 82(5), 1939-1948 (2014-02-26)
Periodontitis is a chronic inflammatory disease with a multifactorial etiology. We investigated whether human major histocompatibility complex (MHC) polymorphisms (6p21.3) are associated with periodontal parameters. Parogene 1 population samples (n = 169) were analyzed with 13,245 single nucleotide polymorphisms (SNPs)
P D Rennert et al.
The Journal of experimental medicine, 184(5), 1999-2006 (1996-11-01)
For more than a decade, the biological roles and the apparent redundancy of the cytokines tumor necrosis factor (TNF) and lymphotoxin (LT) have been debated. LT alpha exists in its soluble form as a homotrimer, which like TNF only binds
Salvador Pastor-Idoate et al.
Clinical ophthalmology (Auckland, N.Z.), 11, 973-981 (2017-06-06)
The objective of this study is to determine the expression and localization of lymphotoxin alpha (LTA) in human retinas and the functionality of one of its polymorphisms rs2229094 (C13R) (T>C), previously associated with proliferative vitreoretinopathy (PVR) development. Total RNA from
Chee Wai Fhu et al.
Blood, 124(19), 2973-2982 (2014-08-21)
It is known that cells within the inflammatory background in classical Hodgkin lymphoma (cHL) provide signals essential for the continual survival of the neoplastic Hodgkin and Reed-Sternberg (HRS) cells. However, the mechanisms underlying the recruitment of this inflammatory infiltrate into

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service