GF94934807
Tin
foil, 6mm disks, thickness 0.025mm, as rolled, 99.99+%
Synonym(s):
Tin, SN000230
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
Sn
CAS Number:
Molecular Weight:
118.71
MDL number:
UNSPSC Code:
12141745
PubChem Substance ID:
NACRES:
NA.23
Assay
≥99.99%
form
foil
manufacturer/tradename
Goodfellow 949-348-07
resistivity
11 μΩ-cm, 20°C
bp
2270 °C (lit.)
mp
231.9 °C (lit.)
density
7.310 g/mL at 25 °C (lit.)
SMILES string
[Sn]
InChI
1S/Sn
InChI key
ATJFFYVFTNAWJD-UHFFFAOYSA-N
Related Categories
General description
For updated SDS information please visit www.goodfellow.com.
Legal Information
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Mercury contamination removed with tin foil.
R Grajower et al.
Operative dentistry, 9(3), 101-104 (1984-01-01)
Britta A Jung et al.
Clinical oral implants research, 22(6), 664-668 (2010-11-04)
To evaluate the necessity of three-dimensional imaging (computed tomography [CT]/cone-beam computed tomography [CBCT]) for paramedian insertion of palatal implants. Lateral radiographs and CBCT scans were performed from 18 human skulls. For lateral cephalometry, the nasal floor (right/left) and the oral
Lis Danielsen et al.
Forensic science international, 134(2-3), 134-141 (2003-07-10)
Previously, electrical injuries have been suggested caused only by the concomitant heat developed during the passage of an electrical current. Recent experimental studies on fully anesthetized pigs and the study of one human case have, however, shown typical electrical alterations.
V G Shlopov et al.
Arkhiv patologii, 46(2), 80-81 (1984-01-01)
The quality of paraffin sections is determined to a considerable degree by the properties of paraffin block such as crystal size and spatial homogeneity. A new temperature regimen of treatment in preparation of paraffin blocks has been developed. After impregnation
K-H Utz et al.
Journal of oral rehabilitation, 29(5), 458-466 (2002-05-25)
In a dentate subject a jaw relation can either be determined in maximum intercuspation and is as such given by the occlusal morphology, or the mandibular position can be allocated according to the centric position of the condyles. For comprehensive
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service