Merck
V800358
Sigma-Aldrich
Silbernitrat
AR, ≥99.9%
Synonym(e):
Salpetersäure Silber(I)-salz
About This Item
Lineare Formel:
AgNO3
CAS-Nummer:
Molekulargewicht:
169.87
EG-Nummer:
MDL-Nummer:
UNSPSC-Code:
12352302
PubChem Substanz-ID:
Qualität
AR
Dampfdichte
5.8 (vs air)
Produktlinie
Vetec
Assay
≥99.9%
Form
solid
mp (Schmelzpunkt)
212 °C (dec.) (lit.)
Anionenspuren
chloride (Cl-): ≤0.0005%
Kationenspuren
Fe: ≤0.0003%
heavy metals (as Pb): ≤0.0005%
SMILES String
[O-][N+]([O-])=O.[Ag+]
InChI
1S/Ag.NO3/c;2-1(3)4/q+1;-1
InChIKey
SQGYOTSLMSWVJD-UHFFFAOYSA-N
Rechtliche Hinweise
Vetec is a trademark of Merck KGaA, Darmstadt, Germany
Signalwort
Danger
Gefahreneinstufungen
Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Ox. Sol. 2 - Repr. 1B - Skin Corr. 1A
Lagerklassenschlüssel
5.1B - Oxidizing hazardous materials
WGK
WGK 3
Flammpunkt (°F)
Not applicable
Flammpunkt (°C)
Not applicable
Die Dokumentenbibliothek aufrufen
Yinqing Zhang et al.
Environmental science & technology, 50(24), 13283-13290 (2016-12-21)
Silver nanowires (AgNWs) are being widely utilized in an increasing number of consumer products, which could release silver to aquatic environments during the use or washing process, and have received growing concerns on their potential risks to bio-organisms and humans.
Radhika P Patil et al.
Nature communications, 11(1), 2923-2923 (2020-06-12)
Porous, nano-architected metals with dimensions down to ~10 nm are predicted to have extraordinarily high strength and stiffness per weight, but have been challenging to fabricate and test experimentally. Here, we use colloidal synthesis to make ~140 nm length and ~15 nm wall
Meice Lu et al.
Materials science & engineering. C, Materials for biological applications, 101, 352-359 (2019-04-29)
Fluorescent carbon quantum dots (CQDs) can be prepared by sonochemistry with low cost and high efficiency. However, ultrasonic synthesis of CQDs with responsive fluorescence towards environmental stimuli remains a big challenge. Herein, we report a simple one-step method to fabricate
Liming Wang et al.
ACS nano, 9(6), 6532-6547 (2015-05-23)
To predict potential medical value or toxicity of nanoparticles (NPs), it is necessary to understand the chemical transformation during intracellular processes of NPs. However, it is a grand challenge to capture a high-resolution image of metallic NPs in a single
Irina Blinova et al.
Environmental science and pollution research international, 20(5), 3456-3463 (2012-11-13)
Although silver nanoparticles (NPs) are increasingly used in various consumer products and produced in industrial scale, information on harmful effects of nanosilver to environmentally relevant organisms is still scarce. This paper studies the adverse effects of silver NPs to two