Merck
Z760994
PCV cell counting tubes
no cap or graduations, for easy read ruler
Synonym(e):
Cell Counting Tubes, PCV Counting Tubes, Tubes for Cell Counting
About This Item
UNSPSC-Code:
41121706
NACRES:
NB.22
Materialien
(PCV)
colorless tube
conical bottom
straight neck
Sterilität
non-sterile
Verpackung
pack of 50 ea
Hersteller/Markenname
TPP 87007
Verfügbarkeit
not available in (available only in North America)
Allgemeine Beschreibung
The packed cell volume (PCV) tube is a quick and reproducible measuring technique of the cell volume that is suitable to monitor cell growth (cell mass).
Leistungsmerkmale und Vorteile
  • Present with or without scale.
  • Fits in microcentrifuges.
  • Caps available if required.
  • Ideal to monitor cell growth
Empfehlung
Produkt-Nr.
Beschreibung
Preisangaben
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)