Merck
Z710970
Rack for glass slides
glass, for 16/32 slides
Synonym(e):
900254
About This Item
UNSPSC-Code:
41122800
NACRES:
MA.01
Materialien
glass
Leistungsmerkmale
Holds 16/32 x slides 76x26 mm
Hersteller/Markenname
WHEATON® 900254
Größe
118x88x70 mm
Allgemeine Beschreibung
Glass rack holds 16 single slides, 31 slides arranged alternately straight across and diagonally, or 32 slides back-to-back.
Rechtliche Hinweise
WHEATON is a registered trademark of Wheaton Industries, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)