Merck
Z710911
Hybridization tube storage rack
Synonym(e):
805015
About This Item
UNSPSC-Code:
41122804
NACRES:
NB.22
Leistungsmerkmale
Holds 6 x hybridization bottles
Hersteller/Markenname
WHEATON® 805015
Allgemeine Beschreibung
  • Acrylic rack for safe transport or laboratory storage. Holds up to six tubes.
Leistungsmerkmale und Vorteile
• Works with 35mm ID WHEATON hybridization bottles• Ideal for transportation and storage• Acrylic construction holds up to 6 bottles
Rechtliche Hinweise
WHEATON is a registered trademark of Wheaton Industries, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)