Z709654
Sartorius mLINE® mechanical pipettor
12 channel, volume 30-300 μL
Synonym(e):
Biohit pipettes, pipettor
Alle Fotos(3)
About This Item
UNSPSC-Code:
41121500
NACRES:
NB.01
Empfohlene Produkte
Leistungsmerkmale
12-channel
electronic: no
Verpackung
pack of 1 ea
Hersteller/Markenname
Sartorius 725240
Messbereich Volumen
30-300 μL
Volumen
300 μL
Allgemeine Beschreibung
Sartorius′s new mechanical pipettor family - the mLINE - offers a unique combination of novel features for mechanical pipetting. Excellent performance, full autoclavability, smooth ergonomics and perfect safety. The mLINE pipettors cover the full volume range of 0.1 to 5000.
- Extremely light pipetting and tip ejection
- Unique safety features
- Stepless volume adjustment and patented volume locking system
- Thermal insulation of internal components to improve accuracy
- Full autoclavability without disassembling
- Easy cleaning, maintenance and calibration
- ID tag and color-coded caps
- Safe-Cone Filters and unique ejection
- Opti-load tip mounting mechanism for even tip sealing and light tip ejection in multichannel pipettors
Rechtliche Hinweise
mLINE is a registered trademark of Sartorius Biohit Liquid Handling Oy
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)