Merck
Z373508
Sigma-Aldrich
Carestream® BioMax® Light Film
Synonym(e):
autoradiography film, xray film
About This Item
UNSPSC-Code:
41105341
NACRES:
NB.22
Größe
7 in. (18 cm) × 9.5 in. (24 cm)
Allgemeine Beschreibung
Doppelter Emulsionsfilm auf transparenter Base. Bietet den klarsten Hintergrund und die höchste Sensitivität für Chemilumineszenz-Detektionssysteme. Auch für Autoradiographie geeignet, jedoch im Vergleich zu X-Omat AR und BioMax MS mit weniger Sensitivität für hochenergetische Isotope und im Vergleich zu BioMax MR mit weniger Sensitivität für mittelhochenergetische Isotope.
Folienblätter ohne Zwischenlagenpapier
Rechtliche Hinweise
BioMax is a registered trademark of Carestream Health, Inc.
Carestream is a registered trademark of Carestream Health, Inc.
Kodak is a registered trademark of Eastman Kodak Company
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Matthias Höllerhage et al.
Scientific reports, 7(1), 11469-11469 (2017-09-15)
α-synuclein-induced neurotoxicity is a core pathogenic event in neurodegenerative synucleinopathies such as Parkinson's disease, dementia with Lewy bodies, or multiple system atrophy. There is currently no disease-modifying therapy available for these diseases. We screened 1,600 FDA-approved drugs for their efficacy
Shaista Naqvi et al.
Proceedings of the National Academy of Sciences of the United States of America, 106(19), 7762-7767 (2009-05-07)
Vitamin deficiency affects up to 50% of the world's population, disproportionately impacting on developing countries where populations endure monotonous, cereal-rich diets. Transgenic plants offer an effective way to increase the vitamin content of staple crops, but thus far it has
Steven C Chen et al.
PloS one, 6(5), e19780-e19780 (2011-05-24)
Although recent publications have linked the molecular events driving facioscapulohumeral muscular dystrophy (FSHD) to expression of the double homeobox transcription factor DUX4, overexpression of FRG1 has been proposed as one alternative causal agent as mice overexpressing FRG1 present with muscular
Sudhakar Raja Subramaniam et al.
Neurobiology of disease, 70, 204-213 (2014-07-13)
Parkinson's disease (PD) is characterized by the progressive degeneration of nigrostriatal dopaminergic neurons leading to motor deficits. The mechanisms underlying the preferential vulnerability of nigrostriatal dopaminergic neurons in PD remain poorly understood. Recent evidence supports a role for mitochondrial dysfunction