Merck
WPB01451WA
Whirl-Pak® Stand-Up Bag
2041 mL capacity(69 oz)
Synonym(e):
Stand-Up Bag, Stand-Up Storage Bag, Whirl-Pak Bag
About This Item
UNSPSC-Code:
24111500
NACRES:
NB.35
Sterilität
sterile
Verpackung
pkg of 250 ea
Hersteller/Markenname
B01451WA (Nasco)
Parameter
-346-180 °F temp. range (-210 to 82 °C)
B × L × Dicke
19 cm × 38 cm × 0.102 mm
Kapazität
2041 mL × 69 oz
2041 mL capacity(69 oz)
Allgemeine Beschreibung
The Stand-Up Bag makes sample handling and testing much easier because it gives users the ability to perform important laboratory functions with both hands, because the bag will stand completely by itself. No rack or other holder is needed to keep the bag upright. When the sample is placed inside, the special gusseted bottom flattens out, allowing the bag to stand up on its own. It works just like a beaker or bottle, however, it costs less, is sterilized, won′t break, and uses only a fraction of storage space as the other containers.
The white, write-on strip makes sample identification easier and more convenient. It′s easy to write on this white strip using an ordinary ballpoint pen, pencil, or felt-tip marker. The paper-like finish virtually eliminates ink smearing, and provides a surface that will accept a variety of writing instruments.
Rechtliche Hinweise
Whirl-pak is a registered trademark of The Aristotle Corporation
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)