WPB01040WA
Whirl-Pak® Thio-Bag®, Waste Water Sampling Bag
100 mL capacity
Synonym(e):
NELAC sample bag, sampling bag
About This Item
Empfohlene Produkte
Sterilität
sterile
Verpackung
pkg of 100 ea
Hersteller/Markenname
Nasco B01040WA
Parameter
-346-180 °F temp. range (-210 to 82 °C)
B × L × Dicke
7.5 cm × 18.5 cm × 0.057 mm
Kapazität
100 mL
100 mL capacity
Verwandte Kategorien
Allgemeine Beschreibung
Capacity is measured in fluid ounces when the bag is closed and tab is folded over three times. Volume and dimensions are approximate; bags should not be used at temperatures above 180° F (82° C). Bags can be frozen to any temperature, even liquid nitrogen temperatures down to -346° F (-210° C) - careful handling required after freezing. All bags are sterilized after manufacturing.
Leistungsmerkmale und Vorteile
- EPA approved
- Self-standing gusseted bag
- Sodium thiosulfate tablet in the bag for chlorine neutralization
- Built-in puncture proof closure
- Post-manufacturing sterilization for results you can trust.
Rechtliche Hinweise
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Kunden haben sich ebenfalls angesehen
Global Trade Item Number
SKU | GTIN |
---|---|
WPB01040WA-100EA | 4061833576359 |