TMO382019-0125
Nalgene® Certified Clean bottles
clear PETG, volume 125 mL, case of 48 ea
Synonym(e):
PETG bottle, certified nalgene bottle
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.43
Empfohlene Produkte
Materialien
clear PETG
Verpackung
case of 48 ea
Hersteller/Markenname
Nalgene 382019-0125
Kapazität
125 mL
Volumen
125 mL
Verwandte Kategorien
Allgemeine Beschreibung
Protect against leakage, breakage and radiation contamination in cleanroom applications with Thermo Scientific Nalgene PETG Certified Clean Containers. Suitable for pharmaceutical and biotechnology applications that require processing and storing bulk intermediates and critical reagents such as vaccine and protein therapeutic preparations. A safe alternative for your lab with no sacrifice of accuracy.
Rechtliche Hinweise
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)